Assay Design



Gene Transcripts

Gene Symbol
Pdzk1
Entrez Gene ID
65144
Gene Name
PDZ domain containing 1
Gene Aliases
Clamp
Chromosome Location
Chr.2: 198965685 - 198999323 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031712.1 NP_113900.1 8 - 9 1590 143
GenBank mRNA AF116896.1 - 8 - 9 1590 143
BC078788.1 - 8 - 9 1582 143

View Full Product Details