Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106459.1 | NP_001099929.1 | 2 - 3 | 513 | 96 |
XM_008761376.1 | XP_008759598.1 | 1 - 2 | 2231 | 96 | |
XM_008761377.1 | XP_008759599.1 | 2 - 3 | 3670 | 96 | |
XM_008761378.1 | XP_008759600.1 | 2 - 3 | 1452 | 96 |