Assay Design
Gene Transcripts
- Gene Symbol
- Clu
- Entrez Gene ID
-
24854
- Gene Name
- clusterin
- Gene Aliases
-
APOJ, CLI, DAG, RATTRPM2B, SGP-2, SGP2, SP-40, SP40, TRPM-2, TRPM2B, Trpm2, Trpmb
- Chromosome Location
-
Chr.15: 42626612 - 42665857 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053021.2 |
NP_444180.2 |
1 - 2 |
53 |
86 |
GenBank mRNA |
M16975.1 |
- |
1 - 2 |
53 |
86 |
View Full Product Details