Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf19
Entrez Gene ID
290300
Gene Name
tumor necrosis factor receptor superfamily, member 19
Gene Aliases
RGD1564996
Chromosome Location
Chr.15: 41249820 - 41316203 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044229.2 NP_001037694.1 2 - 3 177 76
GenBank mRNA DQ466087.1 - 2 - 3 177 76

View Full Product Details