Assay Design



Gene Transcripts

Gene Symbol
Fbxo8
Entrez Gene ID
306436
Gene Name
F-box protein 8
Gene Aliases
-
Chromosome Location
Chr.16: 37131321 - 37177033 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012050.1 NP_001012050.1 2 - 3 563 150
XM_006253091.2 XP_006253153.1 2 - 3 366 150
GenBank mRNA BC088452.1 - 2 - 3 563 150
FQ232328.1 - 2 - 3 620 150

View Full Product Details