Assay Design



Gene Transcripts

Gene Symbol
Cdca7l
Entrez Gene ID
619566
Gene Name
cell division cycle associated 7 like
Gene Aliases
-
Chromosome Location
Chr.6: 145740035 - 145785607 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034953.1 NP_001030125.1 3 - 4 332 94
GenBank mRNA BC098734.1 - 3 - 4 332 94

View Full Product Details