Assay Design



Gene Transcripts

Gene Symbol
Reep6
Entrez Gene ID
362835
Gene Name
receptor accessory protein 6
Gene Aliases
Dp1l1
Chromosome Location
Chr.7: 12240059 - 12246999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013218.1 NP_001013236.1 3 - 4 375 66
XM_006240967.2 XP_006241029.1 3 - 4 645 66
GenBank mRNA BC083830.1 - 3 - 4 375 66

View Full Product Details