Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014051.1 | NP_001014073.1 | 3 - 4 | 189 | 90 |
XM_006235276.2 | XP_006235338.1 | 3 - 4 | 960 | 90 | |
XM_008762331.1 | XP_008760553.1 | 4 - 5 | 244 | 90 | |
XM_008762332.1 | XP_008760554.1 | 4 - 5 | 244 | 90 | |
XM_008762333.1 | XP_008760555.1 | 4 - 5 | 244 | 90 | |
GenBank mRNA | BC082105.1 | - | 3 - 4 | 189 | 90 |