Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053824.2 | NP_446276.1 | 8 - 9 | 698 | 88 |
XM_006235220.2 | XP_006235282.1 | 7 - 8 | 758 | 88 | |
XM_006235221.2 | XP_006235283.1 | 7 - 8 | 799 | 88 | |
GenBank mRNA | BC091130.1 | - | 8 - 9 | 698 | 88 |
L15618.1 | - | - | 1315 | 88 |