Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107789.1 | NP_001101259.1 | 8 - 9 | 922 | 61 |
XM_006235268.1 | XP_006235330.1 | 8 - 9 | 933 | 61 | |
XM_006235269.1 | XP_006235331.1 | 8 - 9 | 933 | 61 | |
XM_008762330.1 | XP_008760552.1 | 8 - 9 | 933 | 61 | |
GenBank mRNA | FQ218565.1 | - | 1 - 2 | 83 | 61 |
FQ220534.1 | - | 1 - 2 | 83 | 61 | |
FQ231155.1 | - | 1 - 2 | 83 | 61 |