Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201655.1 | NP_970617.1 | 2 - 3 | 409 | 70 |
XM_008772274.1 | XP_008770496.1 | 2 - 3 | 686 | 70 | |
XM_008772275.1 | XP_008770497.1 | 2 - 3 | 686 | 70 | |
XM_008772276.1 | XP_008770498.1 | 2 - 3 | 686 | 70 |