Assay Design



Gene Transcripts

Gene Symbol
Ces5a
Entrez Gene ID
307660
Gene Name
carboxylesterase 5A
Gene Aliases
Ces5, Ces7
Chromosome Location
Chr.19: 12097529 - 12136579 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012056.1 NP_001012056.1 5 - 6 814 82
GenBank mRNA AF479659.1 - 5 - 6 814 82

View Full Product Details