Assay Design



Gene Transcripts

Gene Symbol
Mmp15
Entrez Gene ID
291848
Gene Name
matrix metallopeptidase 15
Gene Aliases
-
Chromosome Location
Chr.19: 10079881 - 10101380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106168.2 NP_001099638.1 2 - 3 846 100

View Full Product Details