Assay Design



Gene Transcripts

Gene Symbol
Ecd
Entrez Gene ID
688968
Gene Name
ecdysoneless homolog (Drosophila)
Gene Aliases
RGD1565023
Chromosome Location
Chr.15: 4399834 - 4431268 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770584.1 XP_008768806.1 1 - 2 308 61
XM_008770585.1 XP_008768807.1 2 - 3 167 61

View Full Product Details