Assay Design



Gene Transcripts

Gene Symbol
Ahrr
Entrez Gene ID
498999
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
-
Chromosome Location
Chr.1: 31603297 - 31698329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024285.1 NP_001019456.1 9 - 10 1030 79
GenBank mRNA AB174900.2 - 9 - 10 1030 79
AY367561.1 - 9 - 10 1030 79

View Full Product Details