Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108372.2 | NP_001101842.2 | 2 - 3 | 344 | 63 |
NM_001141929.1 | NP_001135401.1 | 2 - 3 | 299 | 63 | |
XM_008770552.1 | XP_008768774.1 | 2 - 3 | 568 | 63 | |
GenBank mRNA | BU670982.1 | - | 2 - 3 | 344 | 63 |
CF110818.1 | - | 1 - 2 | 42 | 63 | |
CK356697.1 | - | 2 - 3 | 299 | 63 |