Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108372.2 | NP_001101842.2 | 4 - 5 | 521 | 80 |
NM_001141929.1 | NP_001135401.1 | 4 - 5 | 476 | 80 | |
XM_008770552.1 | XP_008768774.1 | 4 - 5 | 745 | 80 | |
GenBank mRNA | CF110818.1 | - | 3 - 4 | 218 | 80 |
CK356697.1 | - | 4 - 5 | 476 | 80 |