Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108372.2 | NP_001101842.2 | 6 - 7 | 677 | 95 |
NM_001141929.1 | NP_001135401.1 | 6 - 7 | 632 | 95 | |
XM_008770552.1 | XP_008768774.1 | 6 - 7 | 901 | 95 | |
GenBank mRNA | CF110818.1 | - | 5 - 6 | 374 | 95 |
CK356697.1 | - | 6 - 7 | 632 | 95 |