Assay Design



Gene Transcripts

Gene Symbol
Irx3
Entrez Gene ID
307721
Gene Name
iroquois homeobox 3
Gene Aliases
-
Chromosome Location
Chr.19: 15838714 - 15842135 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255191.2 XP_006255253.1 2 - 3 2105 134

View Full Product Details