Assay Design



Gene Transcripts

Gene Symbol
Add3
Entrez Gene ID
25230
Gene Name
adducin 3 (gamma)
Gene Aliases
-
Chromosome Location
Chr.1: 273854189 - 273961982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164103.1 NP_001157575.1 12 - 13 1825 160
NM_031552.2 NP_113740.2 12 - 13 1825 160
XM_006231599.2 XP_006231661.1 12 - 13 1678 160
XM_006231600.2 XP_006231662.1 13 - 14 1925 160
XM_006231601.2 XP_006231663.1 12 - 13 2524 160
XM_006231602.2 XP_006231664.1 12 - 13 2526 160
XM_008760510.1 XP_008758732.1 12 - 13 1668 160
XM_008760511.1 XP_008758733.1 12 - 13 2523 160
GenBank mRNA CO402732.1 - 3 - 4 311 160
U35775.1 - - 1742 160

View Full Product Details