Assay Design



Gene Transcripts

Gene Symbol
Add3
Entrez Gene ID
25230
Gene Name
adducin 3 (gamma)
Gene Aliases
-
Chromosome Location
Chr.1: 273854189 - 273961982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164103.1 NP_001157575.1 4 - 5 701 112
NM_031552.2 NP_113740.2 4 - 5 701 112
XM_006231599.2 XP_006231661.1 4 - 5 554 112
XM_006231600.2 XP_006231662.1 5 - 6 801 112
XM_006231601.2 XP_006231663.1 4 - 5 1400 112
XM_006231602.2 XP_006231664.1 4 - 5 1402 112
XM_008760510.1 XP_008758732.1 4 - 5 544 112
XM_008760511.1 XP_008758733.1 4 - 5 1399 112
GenBank mRNA FM072500.1 - 4 - 5 536 112

View Full Product Details