Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106360.2 | NP_001099830.1 | 3 - 4 | 326 | 75 |
GenBank mRNA | BC166889.1 | - | 3 - 4 | 326 | 75 |
BC166905.1 | - | 3 - 4 | 326 | 75 | |
BC166906.1 | - | 3 - 4 | 326 | 75 | |
FQ217125.1 | - | 3 - 4 | 320 | 75 | |
FQ217646.1 | - | 3 - 4 | 329 | 75 | |
FQ223847.1 | - | 3 - 4 | 341 | 75 |