Assay Design



Gene Transcripts

Gene Symbol
Cacnb3
Entrez Gene ID
25297
Gene Name
calcium channel, voltage-dependent, beta 3 subunit
Gene Aliases
CACH3B
Chromosome Location
Chr.7: 140311140 - 140324902 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012828.2 NP_036960.1 5 - 6 563 93
XM_006257329.2 XP_006257391.1 5 - 6 635 93
XM_006257330.2 XP_006257392.1 5 - 6 1044 93
GenBank mRNA M88751.1 - 5 - 6 567 93

View Full Product Details