Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012850.1 | NP_036982.1 | 11 - 12 | 1129 | 75 |
GenBank mRNA | AF122055.1 | - | 11 - 12 | 1102 | 75 |
AF409106.1 | - | 2 - 3 | 301 | 75 | |
L01407.1 | - | 11 - 12 | 1129 | 75 |