Assay Design



Gene Transcripts

Gene Symbol
Mog
Entrez Gene ID
24558
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
-
Chromosome Location
Chr.20: 2003575 - 2014284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022668.2 NP_073159.2 1 - 2 173 61
XM_006255867.2 XP_006255929.1 1 - 2 646 61
XM_008772687.1 XP_008770909.1 1 - 2 650 61
GenBank mRNA BC087717.1 - 1 - 2 173 61
FQ214199.1 - 1 - 2 153 61
M99485.1 - 1 - 2 173 61

View Full Product Details