Assay Design



Gene Transcripts

Gene Symbol
Capn6
Entrez Gene ID
83685
Gene Name
calpain 6
Gene Aliases
-
Chromosome Location
Chr.X: 115049113 - 115073824 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031808.1 NP_113996.1 1 - 2 145 162
GenBank mRNA AF067793.1 - 1 - 2 145 162

View Full Product Details