Assay Design



Gene Transcripts

Gene Symbol
Dvl1
Entrez Gene ID
83721
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
dvl-1
Chromosome Location
Chr.5: 173295955 - 173308014 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031820.1 NP_114008.1 14 - 15 1712 128
XM_006239598.2 XP_006239660.1 14 - 15 1951 128
XM_008764388.1 XP_008762610.1 13 - 14 2114 128
XM_008764389.1 XP_008762611.1 13 - 14 2039 128
XM_008764391.1 XP_008762613.1 14 - 15 1815 128
GenBank mRNA AF143545.2 - 14 - 15 1712 128
AF143546.2 - 14 - 15 1712 128
BC062070.1 - 10 - 11 1974 128
BC160920.1 - 7 - 8 786 128

View Full Product Details