Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133401.1 | NP_596892.1 | 16 - 17 | 2122 | 79 |
XM_006235994.2 | XP_006236056.1 | 18 - 19 | 2947 | 79 | |
GenBank mRNA | AF257746.1 | - | 16 - 17 | 2122 | 79 |
AF286167.2 | - | 15 - 16 | 2037 | 79 | |
AY582535.1 | - | 15 - 16 | 2037 | 79 | |
S66618.1 | - | 3 - 4 | 487 | 79 |