Assay Design
Gene Transcripts
- Gene Symbol
- Clca4
- Entrez Gene ID
-
362053
- Gene Name
- chloride channel accessory 4
- Gene Aliases
-
Clca6, Prp3
- Chromosome Location
-
Chr.2: 250843165 - 250864060 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201419.1 |
NP_958822.1 |
7 - 8 |
1026 |
112 |
GenBank mRNA |
AY333961.1 |
- |
7 - 8 |
1026 |
112 |
View Full Product Details