Assay Design



Gene Transcripts

Gene Symbol
Slc6a12
Entrez Gene ID
50676
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 12
Gene Aliases
BGT1, Gat1, RNU28927, VGAT
Chromosome Location
Chr.4: 153921199 - 153941333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017335.1 NP_059031.1 13 - 14 1875 134
GenBank mRNA FQ229573.1 - - 1399 134
U28927.1 - 13 - 14 1875 134

View Full Product Details