Assay Design



Gene Transcripts

Gene Symbol
Chrna3
Entrez Gene ID
25101
Gene Name
cholinergic receptor, nicotinic, alpha 3 (neuronal)
Gene Aliases
-
Chromosome Location
Chr.8: 59594007 - 59607122 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052805.2 NP_434692.2 5 - 6 1364 65
GenBank mRNA AY574254.1 - 5 - 6 1364 65
L31621.1 - 5 - 6 1440 65
X03440.1 - 5 - 6 1514 65

View Full Product Details