Assay Design
Gene Transcripts
- Gene Symbol
- Klf15
- Entrez Gene ID
-
85497
- Gene Name
- Kruppel-like factor 15
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 122365092 - 122377690 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053536.1 |
NP_445988.1 |
1 - 2 |
53 |
57 |
GenBank mRNA |
AB020759.1 |
- |
1 - 2 |
53 |
57 |
View Full Product Details