Assay Design
Gene Transcripts
- Gene Symbol
- Kremen1
- Entrez Gene ID
-
114107
- Gene Name
- kringle containing transmembrane protein 1
- Gene Aliases
-
Kremen
- Chromosome Location
-
Chr.14: 85441209 - 85503661 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053649.1 |
NP_446101.1 |
5 - 6 |
642 |
125 |
GenBank mRNA |
AB065090.1 |
- |
5 - 6 |
642 |
125 |
View Full Product Details