Assay Design



Gene Transcripts

Gene Symbol
Eif2b3
Entrez Gene ID
171145
Gene Name
eukaryotic translation initiation factor 2B, subunit 3
Gene Aliases
-
Chromosome Location
Chr.5: 135882955 - 135949276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133609.2 NP_598293.2 10 - 11 1316 80
XM_006238630.2 XP_006238692.1 10 - 11 1287 80
GenBank mRNA BC072507.1 - 10 - 11 1316 80
U38253.1 - 10 - 11 80

View Full Product Details