Assay Design



Gene Transcripts

Gene Symbol
Mat2a
Entrez Gene ID
171347
Gene Name
methionine adenosyltransferase II, alpha
Gene Aliases
Sams2
Chromosome Location
Chr.4: 100297478 - 100303047 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134351.1 NP_599178.1 7 - 8 1025 127
GenBank mRNA BC062394.1 - - 1058 127
J05571.1 - 7 - 8 1025 127

View Full Product Details