Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017136.2 | NP_058832.1 | 9 - 10 | 1504 | 104 |
GenBank mRNA | BC097330.1 | - | 9 - 10 | 1504 | 104 |
D37920.1 | - | 9 - 10 | 1635 | 104 | |
FQ210281.1 | - | 1 - 2 | 82 | 104 |