Assay Design
Gene Transcripts
- Gene Symbol
- Pitpnb
- Entrez Gene ID
-
114561
- Gene Name
- phosphatidylinositol transfer protein, beta
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 51279194 - 51341731 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053742.1 |
NP_446194.1 |
10 - 11 |
797 |
129 |
GenBank mRNA |
D21132.1 |
- |
10 - 11 |
797 |
129 |
View Full Product Details