Assay Design
Gene Transcripts
- Gene Symbol
- Dcbld2
- Entrez Gene ID
-
155696
- Gene Name
- discoidin, CUB and LCCL domain containing 2
- Gene Aliases
-
Esdn
- Chromosome Location
-
Chr.11: 44241111 - 44292884 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130419.1 |
NP_569103.1 |
4 - 5 |
630 |
144 |
GenBank mRNA |
AF387549.1 |
- |
4 - 5 |
630 |
144 |
View Full Product Details