Assay Design
Gene Transcripts
- Gene Symbol
- Rap2b
- Entrez Gene ID
-
170923
- Gene Name
- RAP2B, member of RAS oncogene family
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 151685251 - 151685802 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_133410.1 |
NP_596901.1 |
1 - 1 |
499 |
103 |
GenBank mRNA |
AF386786.1 |
- |
1 - 1 |
499 |
103 |
View Full Product Details