Assay Design



Gene Transcripts

Gene Symbol
Golgb1
Entrez Gene ID
192243
Gene Name
golgin B1
Gene Aliases
-
Chromosome Location
Chr.11: 66761645 - 66820989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138885.1 NP_620240.1 14 - 15 8749 121
XM_006248379.2 XP_006248441.1 15 - 16 10742 121
XM_006248380.2 XP_006248442.1 14 - 15 10590 121
XM_008768698.1 XP_008766920.1 14 - 15 9935 121
XM_008768699.1 XP_008766921.1 14 - 15 9920 121
XM_008768700.1 XP_008766922.1 14 - 15 9917 121
XM_008768701.1 XP_008766923.1 13 - 14 9812 121
XM_008768702.1 XP_008766924.1 12 - 13 8461 121
GenBank mRNA D25543.1 - 14 - 15 8749 121

View Full Product Details