Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139253.1 | NP_640346.1 | 1 - 2 | 67 | 51 |
XM_006239710.2 | XP_006239772.1 | 1 - 2 | 106 | 51 | |
GenBank mRNA | AF475282.1 | - | 1 - 2 | 67 | 51 |
BC071179.1 | - | 1 - 2 | 61 | 51 |