Assay Design



Gene Transcripts

Gene Symbol
Mapkapk2
Entrez Gene ID
289014
Gene Name
mitogen-activated protein kinase-activated protein kinase 2
Gene Aliases
-
Chromosome Location
Chr.13: 47785161 - 47871684 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178102.2 NP_835203.1 8 - 9 1149 99
GenBank mRNA AY197741.1 - 8 - 9 1149 99
BC062048.1 - 8 - 9 1115 99

View Full Product Details