Assay Design



Gene Transcripts

Gene Symbol
Sectm1b
Entrez Gene ID
287884
Gene Name
secreted and transmembrane 1B
Gene Aliases
Sectm1
Chromosome Location
Chr.10: 110251363 - 110260565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199082.2 NP_954513.2 3 - 4 514 93
XM_006247882.2 XP_006247944.1 3 - 4 607 93
XM_006247884.2 XP_006247946.1 4 - 5 1432 93
GenBank mRNA BC061794.1 - 3 - 4 93
BQ201063.1 - 1 - 2 93
DY309177.1 - 2 - 3 602 93

View Full Product Details