Assay Design
Gene Transcripts
- Gene Symbol
- Bre
- Entrez Gene ID
-
362704
- Gene Name
- brain and reproductive organ-expressed (TNFRSF1A modulator)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 25666654 - 26051267 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199270.1 |
NP_954891.1 |
1 - 2 |
53 |
100 |
GenBank mRNA |
BC061573.1 |
- |
1 - 2 |
53 |
100 |
View Full Product Details