Assay Design
Gene Transcripts
- Gene Symbol
- Sycp3
- Entrez Gene ID
-
25561
- Gene Name
- synaptonemal complex protein 3
- Gene Aliases
-
RNASCP3
- Chromosome Location
-
Chr.7: 29029887 - 29044344 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_013041.1 |
NP_037173.1 |
7 - 8 |
678 |
123 |
GenBank mRNA |
X75785.1 |
- |
7 - 8 |
678 |
123 |
View Full Product Details