Assay Design



Gene Transcripts

Gene Symbol
Lin7a
Entrez Gene ID
85327
Gene Name
lin-7 homolog a (C. elegans)
Gene Aliases
Veli1
Chromosome Location
Chr.7: 49429613 - 49627835 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053514.1 NP_445966.1 - 40 86
GenBank mRNA AF090134.1 - - 40 86
AF090135.1 - - 40 86

View Full Product Details