Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053738.1 | NP_446190.1 | 1 - 2 | 457 | 68 |
XM_006241395.2 | XP_006241457.1 | 1 - 2 | 472 | 68 | |
GenBank mRNA | AY030278.1 | - | 1 - 2 | 457 | 68 |
BC072473.1 | - | 1 - 2 | 283 | 68 |