Assay Design



Gene Transcripts

Gene Symbol
Mapkapk3
Entrez Gene ID
315994
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
MAPKAP-K3, MK-3
Chromosome Location
Chr.8: 116012129 - 116046021 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012127.1 NP_001012127.1 2 - 3 507 113
XM_006243776.2 XP_006243838.1 2 - 3 597 113
XM_008766531.1 XP_008764753.1 2 - 3 606 113
GenBank mRNA BC081974.1 - 2 - 3 507 113

View Full Product Details