Assay Design



Gene Transcripts

Gene Symbol
Cers6
Entrez Gene ID
366065
Gene Name
ceramide synthase 6
Gene Aliases
Lass6, RGD1561599
Chromosome Location
Chr.3: 55094234 - 55338370 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234308.2 XP_006234370.1 1 - 2 701 66
XM_345363.7 XP_345364.5 1 - 2 695 66

View Full Product Details