Assay Design
Gene Transcripts
- Gene Symbol
- Cers6
- Entrez Gene ID
-
366065
- Gene Name
- ceramide synthase 6
- Gene Aliases
-
Lass6, RGD1561599
- Chromosome Location
-
Chr.3: 55094234 - 55338370 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_345363.7 |
XP_345364.5 |
9 - 10 |
1536 |
116 |
View Full Product Details